Aminomethansulfonsyre 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

155740500 155745000
ACRO155740500EA 320 DKK
ACRO155740500 ACRO155745000
Aminomethansulfonsyre 97%
Aminomethansulfonsyre
Formel: CH₅NO₃S
Smeltepunkt: 180…185 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008124
CAS-nummer: 13881-91-9
EINECS: 237-649-6
UN nummer: 2585
ADR: 8,III

Bestil nu

Specifikationer Testresultater

Appearance White Powder
Infrared spectrum Conforms
Melting point 180 to 185 °C
Titration with NaOH ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR