Tris-(hydroxymethyl) aminomethan (TRIS)
Leverandør: Roth Carl
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
4855.2 4855.1 4855.3
ROTH4855.2EA 0 DKK
ROTH4855.2 ROTH4855.1 ROTH4855.3
Tris-(hydroxymethyl) aminomethan (TRIS)
Tris-(hydroxymethyl) aminomethan (TRIS)
Formel: NH₂C(CH₂OH)₃
Kogepunkt: 357 °C (1013 hPa)
Smeltepunkt: 169…172 °C
Massefylde: 1,328 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004679
CAS-nummer: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM