Precision balances, Pioneer PX series
Leverandør: OHAUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30429812 30429805 30430015 30429814 30429807 30430021 30429816 30429809 30430024 30619382 30619383 30619384 30619385 30714491 30619386 30868072 30868073
611-3842EA 9520 DKK
611-3842 611-3843 611-3844 611-3848 611-3849 611-3850 611-3854 611-3855 611-3856 611-5039 611-5040 611-5042 611-5044 611-5093 611-5091 OHAU30868072 OHAU30868073
Precision balances, Pioneer PX series
Vægte
The Pioneer PX series combines essential weighing functionality with performance, offering high accuracy and repeatability for applications in laboratory, industrial and education settings. PX models are economically priced, and intuitively designed for intelligent operation. Their durable design has a cast metal lower housing, metal sub-pan and stainless steel weighing pan for long-term use.
  • Large viewing angle with bright backlit LCD
  • 2-line display provides additional information or guidance prompts to user
  • A static removal bar (permanent static electric dissipative ABS) provides a convenient grounding location (1 mg precision models)
  • Four quick keys enable easy operation of multiple weighing modes
  • Applications include basic weighing, part counting, percent weighing, dynamic weighing and density determination
  • RS232 and USB ports allow easy connection to PCs and PLCs
Precision models have a real time clock with GLP/GMP data integrated, calibration lock, user-selectable environmental filters and brightness settings. The models feature user-selectable span calibration points, software lockout and reset menu, user-selectable communication settings and data print options, user-definable project and user IDs, software overload/underload indicator, stability indicator and four operating languages and are equipped with a security lock to prevent theft.