Analytical balances, Pioneer PX series
Leverandør: OHAUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30429810 30429803 30430010 30429811 30429804 30430013
611-3836EA 13530 DKK
611-3836 611-3837 611-3838 611-3839 611-3840 611-3841
Analytical balances, Pioneer PX series
Vægte
The Pioneer PX series combines essential weighing functionality with performance, offering high accuracy and repeatability for applications in laboratory, industrial and education settings. PX models are economically priced, and intuitively designed for intelligent operation. Their durable design has a cast metal lower housing, metal sub-pan and stainless steel weighing pan for long-term use.
  • Large viewing angle with bright backlit LCD
  • 2-line display provides additional information or guidance prompts to user
  • A static removal bar (permanent static electric dissipative ABS) provides a convenient grounding location
  • Four quick keys enable easy operation of multiple weighing modes
  • Applications include basic weighing, part counting, percent weighing, dynamic weighing and density determination
  • RS232 and USB ports allow easy connection to PCs and PLCs
Analytical models have a real time clock with GLP/GMP data integrated, calibration lock, user-selectable environmental filters and brightness settings. The models feature user-selectable span calibration points, software lockout and reset menu, user-selectable communication settings and data print options, user-definable project and user IDs, software overload/underload indicator, stability indicator, four operating languages and are equipped with a security lock to prevent theft.