1-Dimethylaminopropan-2-ol 99%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 99%
CAS No.: 108-16-7
Molecular Formula: C5H13NO
Formula Weight: 103.16
Flash Point: 35°(95°F)
Refractive Index: 1.4195
Odour: Amine-like
pH: 11
Melting Point: -85°
Boiling Point: 123-126°
UN No.: 2734
MDL No.: MFCD00004532
Formel:
C₅H₁₃NO Kogepunkt: 123…126 °C Smeltepunkt: –85 °C Massefylde: 0,85 Flammepunkt: 35 °C (95 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004532 CAS-nummer: 108-16-7 EINECS: 203-556-4 UN nummer: 2734 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...