N,N-Dimethylhexylamin 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
314740050
314740250
ACRO314740050EA
516
DKK
ACRO314740050
ACRO314740250
N,N-Dimethylhexylamin 99%
N,N-Dimethylhexylamin
Formel:
C₈H₁₉N Kogepunkt: 146…150 °C (760 mmHg) Massefylde: 0,744 g/cm³ Flammepunkt: 33 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00009522 CAS-nummer: 4385-04-0 EINECS: 224-491-8 UN nummer: 2734 ADR: 8,II |
Specifikationer Testresultater
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4126 to 1.4146 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...