N,N-Dimethylformamid dipropyl acetal ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1,1-Dipropoxytrimethylamine, N,n-dimethylformamidedipropylacetal, 1,1-Dipropoxy-N,N-dimethylmethylamine

L00643.09 L00643.18
L00643.09EA 523 DKK
L00643.09 L00643.18
N,N-Dimethylformamid dipropyl acetal ≥97%
N,N-Dimethylformamid dipropyl acetal

Purity: 97% CAS No.: 6006-65-1 Molecular Formula: C9H21NO2 Formula Weight: 175.27 Flash Point: 46°(115°F) Refractive Index: 1.4088 Boiling Point: 67-68°/14mm Beilstein Reference: 1098524 UN No.: 1993 MDL No.: MFCD00009374

Formel: C₉H₂₁NO₂
Molekylvægt: 175.27 g/mol
Kogepunkt: 67…68 °C (14 mmHg)
Massefylde: 0.854
Flammepunkt: 46 °C (115 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009374
CAS-nummer: 6006-65-1
EINECS: 227-855-4
ADR: 3 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR