Iodacetonitril ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

L03634.09 L03634.18
L03634.09EA 694 DKK
L03634.09 L03634.18
Iodacetonitril ≥97%
Iodacetonitril

Purity: 97% CAS No.: 624-75-9 Molecular Formula: C2H2IN Formula Weight: 166.95 Flash Point: 86°(186°F) Refractive Index: 1.5740 Boiling Point: 70-72°/10mm Beilstein Reference: 1734628 UN No.: 2922 MDL No.: MFCD00001886

Formel: C₂H₂IN
Kogepunkt: 70…72 °C (10 mmHg)
Massefylde: 2,306
Flammepunkt: 86 °C (186 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001886
CAS-nummer: 624-75-9
EINECS: 210-860-0
UN nummer: 2922
ADR: 8,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR