Natriumtrifluormethansulphonat ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Trifluoromethanesulphonic acid sodium salt
T1550-25G T1550-5G
TCIAT1550-25GEA 887 DKK
TCIAT1550-25G TCIAT1550-5G
Natriumtrifluormethansulphonat ≥98.0% (by titrimetric analysis)
Natriumtrifluormethansulphonat
Formel: CF₃NaO₃S
Molekylvægt: 172.06 g/mol
MDL Number: MFCD00061607
CAS-nummer: 2926-30-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91384b45923b225b013f8e8f8809d9b3b58414e3&vid=91384b45923b225b013f8e8f8809d9b3b58414e3&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %
Solubility in Water very faint turbidity