Natriumtrifluormethansulphonat ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Trifluoromethanesulphonic acid sodium salt
B20776.06
B20776.22
B20776.14
B20776.06EA
684
DKK
B20776.06
B20776.22
B20776.14
Natriumtrifluormethansulphonat ≥98%
Natriumtrifluormethansulphonat
Purity: 98%
CAS No.: 2926-30-9
Molecular Formula: CF3SO3Na
Formula Weight: 172.06
Melting Point: 251-255°
Beilstein Reference: 3728797
Formel:
CF₃NaO₃S Smeltepunkt: 251…255 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00061607 CAS-nummer: 2926-30-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...