VWR® Advanced, Bouffant Caps, Latex-Free
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
113-8890EA 829 DKK
113-8890 113-8894 113-8893 113-8891 113-8889 113-8892
VWR® Advanced, Bouffant Caps, Latex-Free
Engangsbeklædning
These disposable caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • Free from natural rubber latex
  • Available in white and blue
Weight: 16 g/m²
Emballering: 100 pieces per bag, 10 bags per case.