4-Aminosmørsyre 99+%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
GABA, γ-Aminobutyric acid
103280250
103281000
103285000
103280025
ACRO103280250EA
202
DKK
ACRO103280250
ACRO103281000
ACRO103285000
ACRO103280025
4-Aminosmørsyre 99+%
4-Aminosmørsyre
Formel:
C₄H₉NO₂ Molekylvægt: 103.12 g/mol Kogepunkt: 248 Smeltepunkt: 197…204 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008226 CAS-nummer: 56-12-2 EINECS: 200-258-6 Merck Index: 13,00429 |
Specifikationer Testresultater
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 197 to 204 °C |
Loss on drying | ≤0.5 % |
Sulfated ash | ≤0.1 % |
UV transmittance | ≥95 % (c=10, 2 N HCl, 430 nm) 1 cm cell |
Titration with HClO4 | ≥99.0 % (On anhydrous substance) |
Chloride (Cl) | ≤400 ppm |
Sulfate (SO4) | ≤500 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...