Methyl3-mercaptopropionat 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-Mercaptopropionic acid methyl ester, Methyl 3-mercaptopropanoate

148870050 148871000 148875000
ACRO148870050EA 237 DKK
ACRO148870050 ACRO148871000 ACRO148875000
Methyl3-mercaptopropionat 98%
Methyl3-mercaptopropionat
Formel: C₄H₈O₂S
Kogepunkt: 54 °C (1013 hPa)
Smeltepunkt: –60 °C
Massefylde: 1,08 g/cm³
Flammepunkt: 60 °C
MDL Number: MFCD00004895
CAS-nummer: 2935-90-2
EINECS: 220-912-4
UN nummer: 3071
ADR: 6.1,II

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4620 to 1.4640 (20°C, 589 nm)
Assay ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR