Dimethylcarbonat for synthesis, Sigma-Aldrich®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig


M-Clarity™ quality level = MQ200

8.03525.0100 8.03525.1000 8.03525.2500
8.03525.0100EA 316 DKK
8.03525.0100 8.03525.1000 8.03525.2500
Dimethylcarbonat for synthesis, Sigma-Aldrich®
Dimethylcarbonat
Formel: C₃H₆O₃
Molekylvægt: 90.08 g/mol
Kogepunkt: 90
Smeltepunkt: 0.5…4.7 °C
Massefylde: 1.07 g/cm³ (20 °C)
Flammepunkt: 18 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008420
CAS-nummer: 616-38-6
EINECS: 210-478-4
UN nummer: 1161
ADR: 3 II
Merck Index: 12,06117

Bestil nu

Specifikationer Testresultater

Solubility 139 g/l (20 °C)
Melting Point 0.5 - 4.7 °C
Molar Mass 90.08 g/mol
Boiling Point 90 °C (1016 hPa)
Vapor Pressure 53 hPa (20 °C)
Flash Point 16.7 °C
Explosion Limit 4.22 - 12.87 %(V)
Density 1.07 g/cm3 (20 °C)
pH (H2O, 20 °C) no data available
Ignition Point 458 °C
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR