Methyllaurat 98+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Dodecylic acid methyl ester, Methyl dodecanoate, Lauric acid methyl ester, Dodecanoic acid methyl ester

215560025 215560100 215560500 215562500
ACRO215560025EA 224 DKK
ACRO215560025 ACRO215560100 ACRO215560500 ACRO215562500
Methyllaurat 98+%
Methyllaurat
Formel: C₁₃H₂₆O₂
Kogepunkt: 262 °C (766 mmHg)
Smeltepunkt: 4…5 °C
Massefylde: 0,87 g/cm³
Flammepunkt: >110 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008966
CAS-nummer: 111-82-0
EINECS: 203-911-3
UN nummer: 3082

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4290 to 1.4340 (20°C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR