Methylbutyrat 99%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Butyric acid methyl ester, Butanoic acid methyl ester
414140050 414141000 414142500
ACRO414140050EA 208 DKK
ACRO414140050 ACRO414141000 ACRO414142500
Methylbutyrat 99%
Methylbutyrat
Formel: C₅H₁₀O₂
Molekylvægt: 102.13 g/mol
Kogepunkt: 102 °C
Smeltepunkt: –84 °C
Massefylde: 0.897 g/cm³
Flammepunkt: 12 °C
MDL Number: MFCD00009391
CAS-nummer: 623-42-7
EINECS: 210-792-1
UN nummer: 1237
ADR: 3 II
Merck Index: 13,06062
Bestil nu
Specifikationer Testresultater
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3850 to 1.3900 (20°C, 589 nm)
Density (20°C) 0.893 to 0.899 g/ml
GC ≥98.5 %
Acid value ≤1 mg KOH/g