Butylbutyrat 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Butyric acid butyl ester

259590010 259590050 259591000
ACRO259590010EA 756 DKK
ACRO259590010 ACRO259590050 ACRO259591000
Butylbutyrat 98%
Butylbutyrat
Formel: C₈H₁₆O₂
Molekylvægt: 144.21 g/mol
Kogepunkt: 166
Smeltepunkt: –92 °C
Massefylde: 0.8675 g/cm³
Flammepunkt: 49 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009450
CAS-nummer: 109-21-7
EINECS: 203-656-8
UN nummer: 1993
ADR: 3 III
Merck Index: 13,01555

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4050 to 1.4070 (20°C, 589 nm)
GC ≥97.5 %
Specific gravity (25°C) 0.866 to 0.869
Acid value ≤1 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR