N,N-Dimethylformamid
Leverandør: Serva
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
DMF
39756.01
39756.02
SERA39756.01EA
238.67
DKK
SERA39756.01
SERA39756.02
N,N-Dimethylformamid
N,N-Dimethylformamid
Formel:
HCON(CH₃)₂ Molekylvægt: 73.09 g/mol Kogepunkt: 153 Smeltepunkt: –61 °C Massefylde: 0.949 g/cm³ (20 °C) Flammepunkt: 57.5 °C (closed cup) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003284 CAS-nummer: 68-12-2 EINECS: 200-679-5 UN nummer: 2265 ADR: 3 III REACH: 01-2119475605-32 Merck Index: 13,03269 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...