N,N-Dimethylformamid, extra dry 99.8%, AcroSeal®

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: DMF

326871000 326870010 326870025
326871000.EA 670 DKK
326871000. 326870010. 326870025.
N,N-Dimethylformamid, extra dry 99.8%, AcroSeal®
N,N-Dimethylformamid
Formel: HCON(CH₃)₂
Molekylvægt: 73.09 g/mol
Kogepunkt: 153
Smeltepunkt: –61 °C
Massefylde: 0.945 g/cm³
Flammepunkt: 58 °C (closed cup)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003284
CAS-nummer: 68-12-2
EINECS: 200-679-5
UN nummer: 2265
ADR: 3 III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 % (Coulometric)
GC ≥99.75 %
Residue after evaporation ≤0.001 %
Acidity (CH3COOH) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR