N,N-Diethylformamid 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Formic acid diethylamide, Formdiethylamide

181371000 181370250 181375000
ACRO181371000EA 841 DKK
ACRO181371000 ACRO181370250 ACRO181375000
N,N-Diethylformamid 99%
N,N-Diethylformamid
Formel: C₅H₁₁NO
Kogepunkt: 176 °C (1013 hPa)
Massefylde: 0,9 g/cm³
Flammepunkt: 60 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003287
CAS-nummer: 617-84-5
EINECS: 210-533-2
UN nummer: 1993
ADR: 3,III

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4330 to 1.4350 (20°C, 589 nm)
GC ≥98.5 %
HPLC ≥92.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR