Butyroylchlorid ≥98%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 98%
CAS No.: 141-75-3
Molecular Formula: CH3CH2CH2COCl
Formula Weight: 106.55
Flash Point: 18°(64°F)
Refractive Index: 1.4120
Odour: Acrid
Melting Point: -89°
Boiling Point: 101-102°
Beilstein Reference: 605395
Merck Reference: 14,1598
UN No.: 2353
MDL No.: MFCD00000752
Formel:
C₄H₇ClO Kogepunkt: 101…102 °C Smeltepunkt: –89 °C Massefylde: 1,026 Flammepunkt: 18 °C (64 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000752 CAS-nummer: 141-75-3 EINECS: 205-498-5 UN nummer: 2353 ADR: 3,II Merck Index: 14,1598 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...