Ethyl acetoacetat ≥99%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Acetoacetic acid ethyl ester, EAA
Purity: 99+%
CAS No.: 141-97-9
Molecular Formula: C6H10O3
Formula Weight: 130.14
Flash Point: 84°(183°F)
Physical Form: Liquid
Density: 1.025
Refractive Index: 1.4190
Appearance: Colourless
Odour: Fruit-like
Melting Point: -43°
Boiling Point: 180-181°
Beilstein Reference: 385838
Merck Reference: 14,3758
MDL No.: MFCD00009199
Formel:
C₆H₁₀O₃ Molekylvægt: 130.14 g/mol Kogepunkt: 180…181 °C Smeltepunkt: –43 °C Massefylde: 1.025 Flammepunkt: 84 °C (183 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00009199 CAS-nummer: 141-97-9 EINECS: 205-516-1 Merck Index: 13,03793 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...