Ethyl acetoacetat 99%, pure
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Acetoacetic acid ethyl ester, EAA
117970025
117970250
117970010
ACRO117970025EA
746
DKK
ACRO117970025
ACRO117970250
ACRO117970010
Ethyl acetoacetat 99%, pure
Ethyl acetoacetat
Formel:
C₆H₁₀O₃ Molekylvægt: 130.14 g/mol Kogepunkt: 180.8 Smeltepunkt: –45 °C Massefylde: 1.03 g/cm³ Flammepunkt: 70 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00009199 CAS-nummer: 141-97-9 EINECS: 205-516-1 Merck Index: 13,03793 |
Specifikationer Testresultater
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4180 to 1.4200 (20°C, 589 nm) |
Water | ≤0.1 % (K.F.) |
GC | ≥98.5 % |
Sulfate (SO4) | ≤0.005 % |
Iron (Fe) | ≤0.001 % |
Sodium (Na) | ≤0.05 % |
Heavy metals (ICP-OES) | ≤5 ppm |
Insoluble matter | ≤0.01 % |
Nitrogen compounds (as N) | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...