3-Chlorpropionsyrechlorid 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

109991000 109995000
ACRO109991000EA 468 DKK
ACRO109991000 ACRO109995000
3-Chlorpropionsyrechlorid 98%
3-Chlorpropionsyrechlorid
Formel: C₃H₄Cl₂O
Molekylvægt: 126.97 g/mol
Kogepunkt: 143…145 °C (760 mmHg)
Smeltepunkt: –32 °C
Massefylde: 1.33 g/cm³
Flammepunkt: 49 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000747
CAS-nummer: 625-36-5
EINECS: 210-890-4
UN nummer: 3489
ADR: 6.1 I

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to dark brown Liquid
Infrared spectrum Conforms
Refractive index 1.4550 to 1.4590 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer About 0.05% 3,5-di-tert-Butylcatechol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR