Trichloracetonitril 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

139370050 139371000 139375000 139370025
ACRO139370050EA 388 DKK
ACRO139370050 ACRO139371000 ACRO139375000 ACRO139370025
Trichloracetonitril 98%
Trichloracetonitril
Formel: C₂Cl₃N
Kogepunkt: 83…84 °C (760 mmHg)
Smeltepunkt: –42 °C
Massefylde: 1,44 g/cm³
Flammepunkt: 195 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00001842
CAS-nummer: 545-06-2
EINECS: 208-885-7
UN nummer: 3276
ADR: 6.1,II
Merck Index: 12,09757

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4420 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR