Dichloracetonitril ≥98%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
A10612.06 A10612.14 A10612.22
A10612.06EA 370 DKK
A10612.06 A10612.14 A10612.22
Dichloracetonitril ≥98%
Dichloracetonitril
Purity: 98+% CAS No.: 3018-12-0 Molecular Formula: C2HCl2N Formula Weight: 109.94 Flash Point: 35°(95°F) Refractive Index: 1.4405 Odour: Pungent Boiling Point: 112-114° Beilstein Reference: 1739029 UN No.: 2920 MDL No.: MFCD00040886
Formel: C₂HCl₂N
Kogepunkt: 112…114 °C
Massefylde: 1,369
Flammepunkt: 35 °C (95 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00040886
CAS-nummer: 3018-12-0
EINECS: 221-159-4
UN nummer: 2920
ADR: 8,II