Octanal ≥98%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Purity: 98%
CAS No.: 124-13-0
Molecular Formula: C8H16O
Formula Weight: 128.22
Flash Point: 56°(133°F)
Refractive Index: 1.4195
Melting Point: 12-15°
Boiling Point: 169-171°
Merck Reference: 14,1766
UN No.: 1191
MDL No.: MFCD00007029
Formel:
C₈H₁₆O Kogepunkt: 169…171 °C Smeltepunkt: 12…15 °C Massefylde: 0,821 Flammepunkt: 56 °C (133 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007029 CAS-nummer: 124-13-0 EINECS: 204-683-8 UN nummer: 1191 ADR: 3,III Merck Index: 14,1766 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...