Bromeddikesyre ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
Leverandør: Merck
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
M-Clarity™ quality level = MQ200
8.02260.0005
8.02260.0250
8.02260.1000
8.02260.0005EA
172
DKK
8.02260.0005
8.02260.0250
8.02260.1000
Bromeddikesyre ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
Bromeddikesyre
Formel:
BrCH₂COOH Molekylvægt: 138.95 g/mol Kogepunkt: 206 Smeltepunkt: 46…49 °C Massefylde: 1.9335 g/cm³ (20 °C) Flammepunkt: >112 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002678 CAS-nummer: 79-08-3 EINECS: 201-175-8 UN nummer: 3425 ADR: 8 II Merck Index: 13,01384 |
Specifikationer Testresultater
Solubility | 93.8 g/l (25 °C) |
Melting Point | 46 - 49 °C |
Molar Mass | 138.94 g/mol |
Boiling Point | 208 °C (1013 hPa) |
Vapor Pressure | 0.158 hPa (25 °C) |
Flash Point |
|
Density | 1.93 g/cm3 (20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...