Bromacetonitril 96%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

A13933.09 A13933.18 A13933.30
A13933.09EA 598 DKK
A13933.09 A13933.18 A13933.30
Bromacetonitril 96%
Bromacetonitril

Purity: 96% CAS No.: 590-17-0 Molecular Formula: BrCH2CN Formula Weight: 119.95 Flash Point: 81°(178°F) Refractive Index: 1.4800 Boiling Point: 150° Beilstein Reference: 956569 UN No.: 3276 MDL No.: MFCD00001884

Formel: BrCNCH₂
Kogepunkt: 150 °C
Smeltepunkt: –36 °C
Massefylde: 1,722
Flammepunkt: 81 °C (178 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001884
CAS-nummer: 590-17-0
EINECS: 209-672-1
UN nummer: 3276
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR