Bromacetonitril 97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
221860050
221860250
221861000
ACRO221860050EA
331
DKK
ACRO221860050
ACRO221860250
ACRO221861000
Bromacetonitril 97%
Bromacetonitril
Formel:
BrCNCH₂ Kogepunkt: 150 °C (1013 hPa) Smeltepunkt: –36 °C Massefylde: 1,72 g/cm³ Flammepunkt: >110 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001884 CAS-nummer: 590-17-0 EINECS: 209-672-1 UN nummer: 2922 ADR: 8,II |
Specifikationer Testresultater
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4780 to 1.4810 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...