1,6-Heptadiyn ≥97%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 97%
CAS No.: 2396-63-6
Molecular Formula: C7H8
Formula Weight: 92.14
Storage Temperature: 2°C to 8°C
Flash Point: 9°(48°F)
Refractive Index: 1.4420
Odour: Recognizable
Boiling Point: 109-111°
Beilstein Reference: 1731756
UN No.: 3295
MDL No.: MFCD00014925
Formel:
C₇H₈ Kogepunkt: 109…111 °C Smeltepunkt: –85 °C Massefylde: 0,810 Flammepunkt: 9 °C (48 °F) Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00014925 CAS-nummer: 2396-63-6 EINECS: 219-253-5 UN nummer: 3295 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...