(S)-(+)-α-Methoxyphenyleddikesyre 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(S)-(+)-α-Methoxyphenylacetic acid, (S)-alpha-Methoxyphenylacetic Acid
224490010
224490050
ACRO224490010EA
982
DKK
ACRO224490010
ACRO224490050
(S)-(+)-α-Methoxyphenyleddikesyre 99%
(S)-(+)-α-Methoxyphenyleddikesyre
Formel:
C₉H₁₀O₃ Molekylvægt: 166.18 g/mol Smeltepunkt: 64…67 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00064216 CAS-nummer: 26164-26-1 EINECS: 247-492-5 |
Specifikationer Testresultater
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 63 to 68 °C |
Specific optical rotation | +145° to +151° (20 °C, 589 nm) (c=0.5, ethanol) |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...