L(-)-Tyrosin, Millipore®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: L-Tyrosine, (S)-(-)-Tyrosine, Tyrosine, H-Tyr-OH

M-Clarity™ quality level = MQ100

1.08371.0025 1.08371.1000 1.08371.9010
1.08371.0025EA 386 DKK
1.08371.0025 1.08371.1000 1.08371.9010
L(-)-Tyrosin, Millipore®
L(-)-Tyrosin
Formel: C₉H₁₁NO₃
Kogepunkt: 351 °C (1013 hPa)
Smeltepunkt: 285 °C
Massefylde: 1,41 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002606
CAS-nummer: 60-18-4
EINECS: 200-460-4
Merck Index: 13,09907

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR