L(+)-Norvalin ≥99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (S)-(+)-Norvaline, L(+)-Norvaline, H-Nva-OH, (S)-2-Aminopentanoic acid

L08658.03 L08658.06
L08658.03EA 469 DKK
L08658.03 L08658.06
L(+)-Norvalin ≥99%
L(+)-Norvalin

Purity: 99% CAS No.: 6600-40-4 Molecular Formula: C5H11NO2 Formula Weight: 117.15 Melting Point: 300° Beilstein Reference: 1721162 Merck Reference: 14,6716 Optical Rotation: +25° (c=10 in 20% HCl) MDL No.: MFCD00064421

Formel: C₅H₁₁NO₂
Molekylvægt: 117.15 g/mol
Kogepunkt: 228
Smeltepunkt: >300 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00064421
CAS-nummer: 6600-40-4
EINECS: 229-543-3
Merck Index: 12,06813

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR