ESD occupational shoes, clogs, C-FIT, Lina
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
974460/35 974460/36 974460/37 974460/38 974460/39 974460/40 974460/41 974460/42
ELTN974460/35EA 403 DKK
ELTN974460/35 ELTN974460/36 ELTN974460/37 ELTN974460/38 ELTN974460/39 ELTN974460/40 ELTN974460/41 ELTN974460/42
ESD occupational shoes, clogs, C-FIT, Lina
Sko og støvler
Leather-free and lightweight clogs made from smooth, tear resistant microfibre upper, an adjustable and foldable heel strap, and a mono-density PU sole (2,5 mm, oil, fuel and abrasion resistant, heat resistant up to approx. 130 °C).
  • Climate regulating, breathable textile lining
  • Full-length inlay sole C-FIT ESD
  • ESD soft fleece insole
  • Anatomically shaped insole and ankle support for improved stability
  • Shock absorber heel
The shoe is tailored to the ergonomics of female feet.
Colour: White
Certificeringer: EN 61340-5-1 (ESD); EN ISO 20347; OB, SRC, A, E, FO