ESD safety shoes, clogs, Mia
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
74440/35 74440/36 74440/37 74440/38 74440/39 74440/40 74440/41 74440/42 74430/35 74430/36 74430/37 74430/38 74430/39 74430/40 74430/41 74430/42
ELTN74440/35EA 432 DKK
ELTN74440/35 ELTN74440/36 ELTN74440/37 ELTN74440/38 ELTN74440/39 ELTN74440/40 ELTN74440/41 ELTN74440/42 ELTN74430/35 ELTN74430/36 ELTN74430/37 ELTN74430/38 ELTN74430/39 ELTN74430/40 ELTN74430/41 ELTN74430/42
ESD safety shoes, clogs, Mia
Sko og støvler
Lightweight anti-static clogs made from smooth, tear resistant microfibre upper, an adjustable heel strap, plus a mono-density PU sole (2,5 mm, oil, fuel and abrasion resistant, heat resistant up to approx. 130 °C).
  • Climate regulating, breathable textile lining
  • Full length inlay sole (C-Fit ESD)
  • ESD soft fleece insole
  • Shock-absorbent heel
  • Protective steel toecap (200 J)
Ergonomically tailored to female feet.
Colour: Black or white
Certificeringer: EN 61340-5-1 (ESD); EN ISO 20345 SB, SRC, A, E, FO.