L(+)-Vinsyre
Leverandør: Roth Carl
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
L-Tartaric Acid, (2R,3R)-(+)-Tartaric acid, Tartaric Acid, (R,R)-(+)-2,3-Dihydroxysuccinic acid, (R,R)-(+)-Tartaric acid, (2R,3R)-2,3-Dihydroxysuccinic acid
K302.1
K302.2
K302.5
ROTHK302.1EA
260.5
DKK
ROTHK302.1
ROTHK302.2
ROTHK302.5
L(+)-Vinsyre
L(+)-Vinsyre
Formel:
C₄H₆O₆ Molekylvægt: 150.09 g/mol Kogepunkt: 399 Smeltepunkt: 168…170 °C Massefylde: 1.759 g/cm³ (20 °C) Flammepunkt: 150 °C (closed cup) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00064207 CAS-nummer: 87-69-4 EINECS: 201-766-0 REACH: 01-2119537204-47 Merck Index: 13,09158 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...