Natriumgluconat ≥99.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Natrium D-gluconat

G0041-25G G0041-500G
TCIAG0041-25GEA 132 DKK
TCIAG0041-25G TCIAG0041-500G
Natriumgluconat ≥99.0% (by titrimetric analysis)
Natriumgluconat
Formel: HOH₂C(CH(OH))₄COONa
Molekylvægt: 218.14 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00064210
CAS-nummer: 527-07-1
EINECS: 208-407-7
Merck Index: 14,04456
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e896b77f0f7bb9db09339d111b36e5f7d11a34e2&vid=e896b77f0f7bb9db09339d111b36e5f7d11a34e2&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 99.0 % (after drying)
Specific rotation [a]20/D +11.0 to +15.0 deg (C=2, H2O)
Solubility in Water almost transparency
Drying loss max. 2.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR