Red IHC chromogen (AP), HIGHDEF®
Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ADI9501400030 ADI9501410030
ENZOADI9501400030EA 1300 DKK
ENZOADI9501400030 ENZOADI9501410030
Red IHC chromogen (AP), HIGHDEF®
Assays Cellulær Assays
Red IHC chromogen (AP) is intended to be used in conjunction with alkaline phosphatase (AP)-based immunostaining or in situ hybridisation systems.
  • Brilliant red colour for AP-activated development
This chromogen substrate system may be used for both automation and manual use.
Leveringsinformation: Substrate buffer and chromogen in an empty mixing bottle.
New Product
Bestil nu
Specifikationer
Environmentally Preferable
Storage conditions Long term
Opbevaringstemperatur 4 °C