Zinksulfat heptahydrat 99.0-103.0% ACS
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
33399.22
33399.36
33399.A4
33399.22EA
295
DKK
33399.22
33399.36
33399.A4
Zinksulfat heptahydrat 99.0-103.0% ACS
Zinksulfat heptahydrat
Purity: 99.0-103.0%
CAS No.: 7446-20-0
Molecular Formula: O4SZn.7H2O
Formula Weight: 287.54 (161.43anhy)
Physical Form: Lump
Density: 1.97
Appearance: Colourless to white
pH: 4.4-6
Melting Point: 100°
Solubility: Soluble in water, glycerol
Merck Reference: 14,10159
UN No.: 3077
MDL No.: MFCD00149894
Formel:
ZnSO₄·7H₂O Smeltepunkt: 100 °C Massefylde: 1,97 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00149894 CAS-nummer: 7446-20-0 EINECS: 231-793-3 UN nummer: 3077 ADR: 9,III Merck Index: 14,10159 |