Zinksulfat monohydrat, EMPROVE® ESSENTIAL Ph. Eur., USP suitable for use as excipient, SAFC®
Leverandør: MERCK PRODUCTION CHEMICALS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
1.08882.5000 1.08882.9025
1.08882.5000EA 23110 DKK
1.08882.5000 1.08882.9025
Zinksulfat monohydrat, EMPROVE® ESSENTIAL Ph. Eur., USP suitable for use as excipient, SAFC®
Zinksulfat monohydrat
Formel: ZnSO₄·1H₂O
Molekylvægt: 179.45 g/mol
Massefylde: 3.2 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149893
CAS-nummer: 7446-19-7
EINECS: 231-793-3
UN nummer: 3077
ADR: 9 III
Merck Index: 13,10213