Chemical resistant gloves, Showa B0700R
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
B0700RL B0700RM B0700RS B0700RXL
112-5968EA 150 DKK
112-5968 112-5969 112-5970 112-5971
Chemical resistant gloves, Showa B0700R
Handsker
These thin, lightweight gloves are ideal for industries where cleanliness is a priority. The flexible PVC coating provides chemical resistance and good grip.
  • Impermeable for working in damp or greasy environments
  • Lint-free, dust-free
  • Fully coated with a scalloped edge and embossed finish for secure grip on slippery objects
  • Non powdered, ‘slip-on’ treatment
  • Seamless liner prevents irritation
  • Designed for easy movement and continuous wear
  • Extended protection on the forearm
Typical applications include: Cleaning, handling pesticides, laboratories, agriculture, healthcare and pharmaceuticals.
Certificeringer: EN ISO 374-1 Type C (KL), EN ISO 374-5, EN 388:2016 (2000X); Cat. III