Lanthan(III)chlorid heptahydrat ≥99%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
A15575.0E A15575.22 A15575.36
A15575.0EEA 11200 DKK
A15575.0E A15575.22 A15575.36
Lanthan(III)chlorid heptahydrat ≥99%
Lanthan(III)chlorid heptahydrat
Purity: 99% CAS No.: 10025-84-0 Molecular Formula: LaCl3.7H2O Formula Weight: 371.37 (245.27anhy) Odour: Odourless Melting Point: 91° dec. MDL No.: MFCD00149756
Formel: LaCl₃·7H₂O
Molekylvægt: 371.37 g/mol
Kogepunkt: ∼860 °C (1013 hPa)
Smeltepunkt: 91 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149756
CAS-nummer: 10025-84-0
EINECS: 233-237-5