Holmiumtribromid, hydrat ≥99.99% (REO, rare earth oxide basis), REacton®

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

13203.09 13203.18
13203.09EA 779 DKK
13203.09 13203.18
Holmiumtribromid, hydrat ≥99.99% (REO, rare earth oxide basis), REacton®
Holmiumtribromid, hydrat

Purity: 99.99% CAS No.: 223911-98-6 Molecular Formula: HoBr3.xH2O Formula Weight: 404.66(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Merck Reference: 14,4727 MDL No.: MFCD00209652

Formel: Br₃Ho·H₂O
Molekylvægt: 422.66 g/mol
Smeltepunkt: 914 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00209652
CAS-nummer: 223911-98-6
EINECS: 237-524-6
Merck Index: 14,4727

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR