Ammoniumjern(III)sulfat dodecahydrat
Leverandør: Roth Carl
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Alum iron, Ammonium iron sulphate dodecahydrate, Ferric ammonium sulphate dodecahydrate, Ammonium iron(III) sulphate dodecahydrate, Iron alum, Ammonium ferric sulphate dodecahydrate
X885.1
X885.2
X885.3
ROTHX885.1EA
175.28
DKK
ROTHX885.1
ROTHX885.2
ROTHX885.3
Ammoniumjern(III)sulfat dodecahydrat
Ammoniumjern(III)sulfat dodecahydrat
Formel:
NH₄Fe(SO₄)₂·12H₂O Molekylvægt: 482.2 g/mol Kogepunkt: 230 Smeltepunkt: 39…41 °C Massefylde: 1.71 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00150004 CAS-nummer: 7783-83-7 EINECS: 233-382-4 Merck Index: 12,00549 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...