Magnesiumsulfat heptahydrat

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: sulphuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt

T888.1 T888.2 T888.5
ROTHT888.1EA 160.36 DKK
ROTHT888.1 ROTHT888.2 ROTHT888.5
Magnesiumsulfat heptahydrat
Magnesiumsulfat heptahydrat
Formel: MgSO₄·7H₂O
Molekylvægt: 246.48 g/mol
Smeltepunkt: 1124
Massefylde: 2.66 g/cm³ (20 °C)
MDL Number: MFCD00149785
CAS-nummer: 10034-99-8
Merck Index: 13,05715

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR