Calciumsulfat dihydrat 98.0-102.0% ACS
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Plaster of Paris
36700.30
36700.A1
36700.30EA
575
DKK
36700.30
36700.A1
Calciumsulfat dihydrat 98.0-102.0% ACS
Calciumsulfat dihydrat
Purity: 98.0-102.0%
CAS No.: 10101-41-4
Molecular Formula: CaSO4.2H2O
Formula Weight: 172.17 (136.14anhy)
Physical Form: Powder
Density: 2.32
Melting Point: 128° -1.5H2O
Boiling Point: 163° -2H2O
Merck Reference: 14,1706
MDL No.: MFCD00149625
Formel:
CaSO₄·2H₂O Kogepunkt: 163 °C (-2H₂O) Massefylde: 2,32 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00149625 CAS-nummer: 10101-41-4 EINECS: 231-900-3 Merck Index: 14,1706 |