Calciumsulfat hemihydrat
Leverandør: GFS CHEMICALS
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3656.0500
3656.2500
GFSC3656.0500EA
0
DKK
GFSC3656.0500
GFSC3656.2500
Calciumsulfat hemihydrat
Calciumsulfat hemihydrat
Formel:
CaSO₄·0.5H₂O Smeltepunkt: > 1400 °C Massefylde: 2,73 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00010912 CAS-nummer: 10034-76-1 EINECS: 231-900-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...