Aluminiumkaliumsulfat dodecahydrat

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Aluminium Potassium sulphate, 12-Hydrate, Aluminum potassium sulphate dodecahydrate, Aluminium potassium bis(sulphate) dodecahydrate, Potassium alum, Alum potassium

P724.1 P724.2
ROTHP724.1EA 458.7 DKK
ROTHP724.1 ROTHP724.2
Aluminiumkaliumsulfat dodecahydrat
Aluminiumkaliumsulfat dodecahydrat
Formel: AlK(SO₄)₂·12H₂O
Molekylvægt: 474.39 g/mol
Smeltepunkt: 92.5
Massefylde: 1.72 g/cm³ (20 °C)
MDL Number: MFCD00149143
CAS-nummer: 7784-24-9
EINECS: 233-141-3
Merck Index: 12,00373

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR