Natriumdihydrogenphosphat dihydrat
Leverandør: Roth Carl
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Phosphoric acid (sodium hydrate), Sodium phosphate monobasic dihydrate, Sodium dihydrogen orthophosphate dihydrate
2370.1 2370.2 2370.3
ROTH2370.1EA 0 DKK
ROTH2370.1 ROTH2370.2 ROTH2370.3
Natriumdihydrogenphosphat dihydrat
Natriumdihydrogenphosphat dihydrat
Formel: NaH₂PO₄·2H₂O
Molekylvægt: 156.01 g/mol
Kogepunkt: ∼650 °C (1013 hPa)
Smeltepunkt: 60
Massefylde: 1.91 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149209
CAS-nummer: 13472-35-0
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806